Invitrogen
Manufacturer Code:PA577720
Catalog # PIPA577720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide CRDPDYGTLIQE corresponding to amino acid residues 27-38 of rat RAMP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: receptor (calcitonin) activity modifying protein 1; Receptor activity-modifying protein 1; Receptor activity-modifying protein 1 Calcitonin Receptor-Like Receptor Activity Modifying Protein 1
Gene Aliases: 9130218E19Rik; Ramp1
UniProt ID: (Rat) Q9JJ74, (Mouse) Q9WTJ5
Entrez Gene ID: (Rat) 58965, (Mouse) 51801
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.