Novus Biologicals
Manufacturer Code:NBP29407820UL
Catalog # NB022716
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 27-117 of human RAMP1 (NP_005846.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcitonin receptor-like receptor activity modifying protein 1; calcitonin receptor-like receptor activity modifying protein 1 Calcitonin-receptor-like receptor activity-modifying protein 1 CRLR activity-modifying protein 1 receptor (calcitonin) activity modifying protein 1 receptor (G protein-coupled) activity modifying protein 1 receptor activity modifying protein 1 receptor activity-modifying protein 1; Calcitonin-receptor-like receptor activity-modifying protein 1; CRLR activity-modifying protein 1; receptor (calcitonin) activity modifying protein 1; receptor (G protein-coupled) activity modifying protein 1; Receptor activity-modifying protein 1
Gene Aliases: RAMP1
UniProt ID: (Human) O60894
Entrez Gene ID: (Human) 10267
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.