R&D Systems
Manufacturer Code:AF6428SP
Catalog # AF6428SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human RAMP1 Cys27-Ser117 Accession # O60894 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: calcitonin receptor-like receptor activity modifying protein 1; calcitonin receptor-like receptor activity modifying protein 1 Calcitonin-receptor-like receptor activity-modifying protein 1 CRLR activity-modifying protein 1 receptor (calcitonin) activity modifying protein 1 receptor (G protein-coupled) activity modifying protein 1 receptor activity modifying protein 1 receptor activity-modifying protein 1; Calcitonin-receptor-like receptor activity-modifying protein 1; CRLR activity-modifying protein 1; receptor (calcitonin) activity modifying protein 1; receptor (G protein-coupled) activity modifying protein 1; Receptor activity-modifying protein 1
Gene Aliases: RAMP1
UniProt ID: (Human) O60894
Entrez Gene ID: (Human) 10267
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.