Novus Biologicals
Manufacturer Code:NBP15293020UL
Catalog # NBP15293020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RALGPS2(Ral GEF with PH domain and SH3 binding motif 2) The peptide sequence was selected from the C terminal of RALGPS2. Peptide sequence YLGIYLSDLTYIDSAYPSTGSILENEQRSNLMNNILRIISDLQQSCEYDI. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dJ595C2.1 FLJ10244 FLJ25604 KIAA0351 Ral GEF with PH domain and SH3 binding motif 2 Ral GEF with PH domain and SH3-binding motif 2 RalA exchange factor RalGPS2 Ral-A exchange factor RalGPS2 ras-specific guanine nucleotide-releasing factor RalGPS2; Ral GEF with PH domain and SH3-binding motif 2; Ral-A exchange factor RalGPS2; RalA exchange factor RalGPS2; Ras-specific guanine nucleotide-releasing factor RalGPS2
Gene Aliases: dJ595C2.1; RALGPS2
UniProt ID: (Human) Q86X27
Entrez Gene ID: (Human) 55103
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.