Invitrogen
Manufacturer Code:PA569173
Catalog # PIPA569173
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human RALGPS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ral GEF with PH domain and SH3 binding motif 1; Ral GEF with PH domain and SH3-binding motif 1; Ral guanine nucleotide exchange factor 2; Ral guanine nucleotide exchange factor RalGPS1A; RalA exchange factor RalGPS1; RalGEF 2; RALGPS1 KIAA0351 RALGEF2; Ras-specific guanine nucleotide-releasing factor RalGPS1
Gene Aliases: 5830418G11Rik; AI853783; AI854138; KIAA0351; mKIAA0351; RalGEF 2; RALGEF2; RALGPS1; RALGPS1A; si:dkey-191c17.1; zgc:165535
UniProt ID: (Human) Q5JS13, (Mouse) A2AR50, (Zebrafish) B0UXH6
Entrez Gene ID: (Human) 9649, (Rabbit) 100357684, (Horse) 100067166, (Bovine) 616166, (Dog) 609271, (Guinea pig) 100728710, (Rat) 100361774, (Mouse) 241308, (Zebrafish) 100101642
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.