Invitrogen
Manufacturer Code:PA5116790
Catalog # PIPA5116790
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human RALGDS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gnds KIAA1308 mKIAA1308 Ral guanine nucleotide dissociation stimulator ral guanine nucleotide exchange factor RALGDS ralGEF Rgds RGF; Ral guanine nucleotide dissociation stimulator; Ral guanine nucleotide exchange factor; RalGDS; RalGEF
Gene Aliases: Gnds; KIAA1308; mKIAA1308; RALGDS; RalGEF; RGDS; RGF
UniProt ID: (Human) Q12967, (Mouse) Q03385, (Rat) Q03386
Entrez Gene ID: (Human) 5900, (Mouse) 19730, (Rat) 29622
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.