Invitrogen
Manufacturer Code:PA571249
Catalog # PIPA571249
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human RALGAPB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p170; p170 {ECO:0000303|PubMed:19520869}; Ral GTPase activating protein non-catalytic beta subunit; Ral GTPase activating protein, beta subunit (non-catalytic); ral GTPase-activating protein beta subunit; Ral GTPase-activating protein subunit beta; ralgapb {ECO:0000303|PubMed:19520869}; zmp:0000000942
Gene Aliases: 9330195P16; AI507211; B230339M05Rik; KIAA1219; mKIAA1219; p170; RALGAPB; RalGAPbeta; RGD1306861
UniProt ID: (Human) Q86X10
Entrez Gene ID: (Human) 57148, (Dog) 612869, (Horse) 100070219, (Rabbit) 100351145, (Mouse) 228850, (Rat) 362257, (Guinea pig) 100731648, (Zebrafish) 559303
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.