Invitrogen
Manufacturer Code:PA538853
Catalog # PIPA538853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human AS250 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 250 kDa substrate of Akt; akt substrate AS250; AS250; Kiaa1272; P220; Ral GTPase activating protein, alpha subunit 2 (catalytic); ral GTPase-activating protein alpha subunit 2; Ral GTPase-activating protein subunit alpha-2; RapGAPalpha2
Gene Aliases: A230067G21Rik; AS250; bA287B20.1; BC053994; C20orf74; dJ1049G11; dJ1049G11.4; KIAA1272; p220; pp250; RALGAPA2; RGC2
UniProt ID: (Human) Q2PPJ7, (Mouse) A3KGS3
Entrez Gene ID: (Human) 57186, (Mouse) 241694
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.