OriGene
Manufacturer Code:TA505880S
Catalog # 50-167-6283
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C4 |
Immunogen | Full length human recombit protein of human RALB produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GTP binding protein); RAS like proto-oncogene B; ras related GTP binding protein B; RAS-like protein B; Ras-related protein Ral-B; v-ral simian leukemia viral oncogene homolog B (ras related); v-ral simian leukemia viral oncogene homolog B (ras related; GTP binding protein)
Gene Aliases: 5730472O18Rik; dRalb; Ral-b; RALB
UniProt ID: (Human) P11234, (Rat) P36860, (Mouse) Q9JIW9
Entrez Gene ID: (Human) 5899, (Dog) 483881, (Rat) 116546, (Mouse) 64143
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.