Invitrogen
Manufacturer Code:PA544575
Catalog # PIPA544575
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human RALB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP binding protein); ras related GTP binding protein B; RAS-like protein B; Ras-related protein Ral-B; v-ral simian leukemia viral oncogene homolog B; v-ral simian leukemia viral oncogene homolog B (ras related); v-ral simian leukemia viral oncogene homolog B (ras related; GTP binding protein)
Gene Aliases: 5730472O18Rik; dRalb; Ral-b; RALB
UniProt ID: (Human) P11234, (Rat) P36860, (Mouse) Q9JIW9
Entrez Gene ID: (Human) 5899, (Horse) 100060199, (Goat) 100861145, (Bovine) 505238, (Dog) 483881, (Rabbit) 100346449, (Rat) 116546, (Guinea pig) 100715093, (Mouse) 64143
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.