OriGene
Manufacturer Code:CF810869
Catalog # 50-167-0354
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9G8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 205-419 of human RAGE produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: advanced glycation end-products receptor; Advanced glycosylation end product-specific receptor; RAGE isoform NtRAGE-delta; RAGE isoform sRAGE-delta; RAGE RAGE-1 RAGE1 STK30; receptor for advanced glycation end-products variant 20; Receptor for advanced glycosylation end products
Gene Aliases: AGER; RAGE; SCARJ1
UniProt ID: (Human) Q9H2X7, (Mouse) Q62151
Entrez Gene ID: (Human) 177, (Mouse) 11596
Molecular Function: cell adhesion molecule immunoglobulin superfamily cell adhesion molecule receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.