Invitrogen
Manufacturer Code:PA578735
Catalog # PIPA578735
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human RAGE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: advanced glycation end-products receptor; Advanced glycosylation end product-specific receptor; Advanced glycosylation end product-specific receptor Receptor for advanced glycosylation end products AGER RAGE; advanced glycosylation end product-specific receptor variant 2; advanced glycosylation end product-specific receptor variant 3; advanced glycosylation end product-specific receptor variant 4; advanced glycosylation end product-specific receptor variant 5; RAGE isoform NtRAGE-delta; RAGE isoform sRAGE-delta; receptor for advanced glycation end-products variant 20; Receptor for advanced glycosylation end products
Gene Aliases: AGER; RAGE; SCARJ1
UniProt ID: (Human) Q9H2X7, (Mouse) Q62151
Entrez Gene ID: (Human) 177, (Rat) 81722, (Mouse) 11596
Molecular Function: cell adhesion molecule immunoglobulin superfamily cell adhesion molecule receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.