Invitrogen
Manufacturer Code:PA579909
Catalog # PIPA579909
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human RAG2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RAG-2; recombinase activating gene 2; recombination activating protein 2; V(D)J recombination-activating protein 2; V(D)J recombination-activating protein 2 RAG-2 RAG2
Gene Aliases: RAG-2; RAG2
UniProt ID: (Human) P55895, (Mouse) P21784
Entrez Gene ID: (Human) 5897, (Rat) 295953, (Mouse) 19374
Molecular Function:
DNA binding protein
endodeoxyribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.