Invitrogen
Manufacturer Code:MA517163
Catalog # PIMA517163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D9 |
Immunogen | Purified recombit fragment of human RAG1 (amino acids: 818-868) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase RAG1; Endonuclease RAG1; RAG-1; recombination activating protein 1; RING finger protein 74; RING-type E3 ubiquitin transferase RAG1; V(D)J recombination-activating protein 1
Gene Aliases: RAG-1; RAG1; RNF74
UniProt ID: (Human) P15918
Entrez Gene ID: (Human) 5896
Molecular Function:
DNA binding protein
endodeoxyribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.