Invitrogen
Manufacturer Code:PA569206
Catalog # PIPA569206
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse RAG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase RAG1; Endonuclease RAG1; RAG-1; recombinase activating gene 1; recombinase activating protein; recombination activating gene-1; recombination activating protein 1; recombination activation protein 1; recombination-activating gene 1; RING-type E3 ubiquitin transferase RAG1; V(D)J recombination-activating protein 1
Gene Aliases: RAG-1; RAG1
UniProt ID: (Pig) Q867B5, (Mouse) P15919
Entrez Gene ID: (Pig) 397506, (Horse) 100049810, (Rat) 84600, (Mouse) 19373
Molecular Function:
DNA binding protein
endodeoxyribonuclease
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.