Invitrogen
Manufacturer Code:PA541738
Catalog # PIPA541738
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human RAE1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homolog of yeast Rae1 (Bharathi) mRNA-associated protein of 41 kDa (Kraemer); migration-inducing gene 14; mRNA export factor; mRNA export factor-like protein; mRNA export protein; mRNA-associated protein MRNP 41; mRNA-binding protein, 41-kD; RAE1 (RNA export 1, S.pombe) homolog; Rae1 protein homolog; RAE1 RNA export 1 homolog; RAE1 RNA export 1 homolog (S. pombe) MIG14 MRNP41 Mnrp41 dJ481F12.3 dJ800J21.1
Gene Aliases: 3230401I12Rik; MNRP; 41; D2Ertd342e; dJ481F12.3; dJ800J21.1; MIG14; MNRP41; MRNP41; RAE1; zgc:56449; zgc:77723
UniProt ID: (Human) P78406, (Bovine) Q5E9A4, (Rat) Q3SWS8, (Mouse) Q8C570, (Zebrafish) Q7ZWF0
Entrez Gene ID: (Human) 8480, (Horse) 100050441, (Bovine) 513080, (Dog) 485942, (Rabbit) 100359017, (Rat) 362281, (Mouse) 66679, (Guinea pig) 100724811, (Zebrafish) 393973
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.