Abnova
Manufacturer Code:PAB6827
Catalog # 89-132-719
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human RAE1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of RAE1.
For Research Use Only
Protein Aliases: dJ481F12.3; dJ800J21.1; FLJ30608; homolog of yeast Rae1 (Bharathi) mRNA-associated protein of 41 kDa (Kraemer); MGC117333; MGC126076; MGC126077; MIG14; migration-inducing gene 14; Mnrp41; mRNA export factor; mRNA export protein; mRNA-associated protein MRNP 41; mRNA-binding protein, 41-kD; MRNP41; Rae1 protein homolog; RAE1 RNA export 1 homolog
Gene Aliases: dJ481F12.3; dJ800J21.1; MIG14; Mnrp41; MRNP41; RAE1
UniProt ID: (Human) P78406
Entrez Gene ID: (Human) 8480
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.