Novus Biologicals
Manufacturer Code:NBP15823320UL
Catalog # NBP15823320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to RAD54B(RAD54 homolog B (S. cerevisiae)) The peptide sequence was selected from the N terminal of RAD54B. Peptide sequence DAVLIVKGKSFILKNLEGKDIGRGIGYKFKELEKIEEGQTLMICGKEIEV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA repair and recombination protein RAD54B; DNA repair and recombination protein RAD54B EC 3.6.1 EC 3.6.4.- fibrinogen silencer binding protein FSBP RAD54 homolog B RAD54 homolog B (S. cerevisiae) RDH54; Fibrinogen silencer-binding protein; RAD54 homolog B
Gene Aliases: FSBP; RAD54B; RDH54
UniProt ID: (Human) O95073
Entrez Gene ID: (Human) 25788
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.