Invitrogen
Manufacturer Code:MA535671
Catalog # PIMA535671
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1183 |
Immunogen | A synthesized peptide derived from human RAD52 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein RAD52 homolog; DNA repair protein RAD52 homolog RAD52 Rad52yh; recombination protein RAD52; rhabdomyosarcoma antigen MU-RMS-40.23
Gene Aliases: RAD52
UniProt ID: (Human) P43351
Entrez Gene ID: (Human) 5893
Molecular Function:
DNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.