Novus Biologicals
Manufacturer Code:NBP29519520UL
Catalog # NB024553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 125-418 of human RAD52 (NP_602296.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein RAD52 homolog; DNA repair protein RAD52 homolog RAD52 (S. cerevisiae) homolog RAD52 homolog (S. cerevisiae) recombination protein RAD52 rhabdomyosarcoma antigen MU-RMS-40.23; recombination protein RAD52; rhabdomyosarcoma antigen MU-RMS-40.23
Gene Aliases: RAD52
UniProt ID: (Human) P43351
Entrez Gene ID: (Human) 5893
Molecular Function: DNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.