Invitrogen
Manufacturer Code:PA518106
Catalog # PIPA518106
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (RGKTFRFEMQRDL) corresponding to the N-terminus amino acids of RAD51C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein RAD51 homolog 3; R51H3; RAD51 homolog C; RAD51-like protein 2; RAD51C RAD51 homolog C (S. cerevisiae) RAD51L2 yeast RAD51 homolog 3 DNA repair protein RAD51 homolog 3 MGC104277 RAD51 homolog C RAD51 homolog C isoform 1; yeast RAD51 homolog 3
Gene Aliases: BROVCA3; FANCO; R51H3; RAD51C; RAD51L2
UniProt ID: (Human) O43502
Entrez Gene ID: (Human) 5889
Molecular Function:
DNA binding protein
DNA strand-pairing protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.