Invitrogen
Manufacturer Code:PA575317
Catalog # PIPA575317
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 201-250 of Human Rad51B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein RAD51 homolog 2; DNA repair protein RAD51 homolog 2 R51H2 RAD51 homolog B Rad51B RAD51-like protein 1 RAD51B RAD51L1 REC2; R51H2; RAD51 homolog B; RAD51-like 1; RAD51-like protein 1; Rad51B; RecA-like protein; recombination repair protein
Gene Aliases: AI553500; mREC2; R51H2; RAD51B; RAD51L1; REC2
UniProt ID: (Human) O15315, (Mouse) O35719
Entrez Gene ID: (Human) 5890, (Mouse) 19363
Molecular Function:
DNA binding protein
DNA strand-pairing protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.