Invitrogen
Manufacturer Code:PA544829
Catalog # PIPA544829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human RAD51L1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein RAD51 homolog 2; R51H2; RAD51 homolog B; RAD51 homolog B (S. cerevisiae) MGC34245 R51H2 RAD51B REC2 hREC2 RAD51L1; RAD51 paralog B; RAD51-like 1; RAD51-like protein 1; Rad51B; RecA-like protein; recombination repair protein
Gene Aliases: AI553500; mREC2; R51H2; RAD51B; RAD51L1; REC2; RGD1560681
UniProt ID: (Human) O15315, (Mouse) O35719
Entrez Gene ID: (Dog) 490746, (Bovine) 617007, (Human) 5890, (Rabbit) 100351948, (Horse) 100053023, (Mouse) 19363, (Rat) 500679, (Guinea pig) 100713111
Molecular Function:
DNA binding protein
DNA strand-pairing protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.