Invitrogen
Manufacturer Code:MA123330
Catalog # PIMA123330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 13E4 |
Immunogen | Recombit human Rad51 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BRCA1/BRCA2-containing complex, subunit 5; BRCC5 HRAD51 HsRad51 HsT16930 RAD51A RECA; DNA repair protein RAD51 homolog 1; HsRAD51; RAD51 homolog A; RecA, E. coli, homolog of; RecA-like protein; recombination protein A
Gene Aliases: BRCC5; FANCR; HRAD51; HsRad51; HsT16930; MRMV2; RAD51; RAD51A; RECA
UniProt ID: (Human) Q06609
Entrez Gene ID: (Human) 5888
Molecular Function: DNA binding protein DNA strand-pairing protein hydrolase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.