Invitrogen
Manufacturer Code:PA534905
Catalog # PIPA534905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 259 of RAD51 (Uniprot IDNo.P25454) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BRCA1/BRCA2-containing complex subunit 5 DNA repair protein RAD51 homolog 1 RAD51 homolog A RecA E. coli homolog of RecA-like protein recombination protein A; DNA repair protein RAD51; recombinase RAD51
Gene Aliases: MUT5; RAD51; YER095W
UniProt ID: (Baker's yeast) P25454
Entrez Gene ID: (Baker's yeast) 856831
Molecular Function:
DNA binding protein
DNA strand-pairing protein
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.