Invitrogen
Manufacturer Code:MA517259
Catalog # PIMA517259
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 20B5 |
Immunogen | Recombit human protein purified from E.coli (His-Rad50) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein RAD50; hRAD50; hRad50 hsRAD50 Mrell RAD50 2 RAD50 homolog (S cerevisiae) RAD50 PEN Rad50l Truncated RAD50 protein DNA repair protein RAD50; mRad50; RAD50 homolog, double strand break repair protein
Gene Aliases: hRad50; Mrell; NBSLD; RAD50; RAD502; Rad50l
UniProt ID: (Human) Q92878, (Rat) Q9JIL8
Entrez Gene ID: (Human) 10111, (Mouse) 19360, (Rat) 64012
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.