Invitrogen
Manufacturer Code:MA123269
Catalog # 01-950-107
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 13B3 |
Immunogen | Amino acids 1-425 of Rad50 expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DNA repair protein RAD50; hRAD50; mRad50; NBSLD RAD50-2 hRad50; RAD50 homolog, double strand break repair protein
Gene Aliases: hRad50; Mrell; NBSLD; RAD50; RAD502; Rad50l
UniProt ID: (Human) Q92878, (Rat) Q9JIL8
Entrez Gene ID: (Human) 10111, (Mouse) 19360, (Rat) 64012
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.