OriGene
Manufacturer Code:CF811221
Catalog # 50-167-0530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A12 |
Immunogen | Human recombit protein fragment corresponding to amino acids 193-458 of human RAD21 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 64-kDa C-terminal product; 64-kDa carboxy-terminal product; 65-kDa carboxy-terminal product; CDLS4 HR21 HRAD21 MCD1 NXP1 SCC1 hHR21; Double-strand-break repair protein rad21 homolog; hHR21; kleisin; Nuclear matrix protein 1; NXP-1; protein involved in DNA double-strand break repair; RAD21 homolog; SCC1 homolog; sister chromatid cohesion 1
Gene Aliases: CDLS4; hHR21; HR21; HRAD21; KIAA0078; MCD1; NXP1; RAD21; SCC1
UniProt ID: (Human) O60216
Entrez Gene ID: (Human) 5885
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.