Invitrogen
Manufacturer Code:PA570233
Catalog # PIPA570233
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of rat RAD21 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 64-kDa C-terminal product; 64-kDa carboxy-terminal product; 65-kDa carboxy-terminal product; Double-strand-break repair protein rad21 homolog; double-strand-break repair protein rad21-like protein; FLJ25655 FLJ40596 HR21 HRAD21 KIAA0078 MCD1 NXP1 SCC1 hHR21 NXP-1 SCC1 homolog double-strand-break repair protein rad21 homolog nuclear matrix protein 1 protein involved in DNA double-strand break repair; hHR21; kleisin; mHR21; Nuclear matrix protein 1; NXP-1; Pokeweed agglutinin-binding protein 29; protein involved in DNA double-strand break repair; PW29; RAD21 homolog; SCC1 homolog; sister chromatid cohesion 1
Gene Aliases: CDLS4; hHR21; HR21; HRAD21; KIAA0078; MCD1; mKIAA0078; NXP1; RAD21; SCC1
UniProt ID: (Human) O60216, (Bovine) Q3SWX9, (Mouse) Q61550
Entrez Gene ID: (Human) 5885, (Rabbit) 100350479, (Dog) 482021, (Bovine) 540966, (Horse) 100065497, (Guinea pig) 100718361, (Rat) 314949, (Mouse) 19357
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.