Abnova
Manufacturer Code:PAB5544
Catalog # 89-131-897
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human RAD21. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of RAD21.
For Research Use Only
Protein Aliases: 64-kDa C-terminal product; 64-kDa carboxy-terminal product; 65-kDa carboxy-terminal product; Double-strand-break repair protein rad21 homolog; FLJ25655; FLJ40596; hHR21; HR21; HRAD21; KIAA0078; kleisin; MCD1; Nuclear matrix protein 1; NXP-1; NXP1; protein involved in DNA double-strand break repair; RAD21 homolog; SCC1; SCC1 homolog; sister chromatid cohesion 1
Gene Aliases: CDLS4; hHR21; HR21; HRAD21; KIAA0078; MCD1; NXP1; RAD21; SCC1
UniProt ID: (Human) O60216
Entrez Gene ID: (Human) 5885
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.