Invitrogen
Manufacturer Code:PA552326
Catalog # PIPA552326
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RAD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cell cycle checkpoint protein Hrad1; Cell cycle checkpoint protein RAD1; checkpoint control protein HRAD1; DNA repair exonuclease rad1 homolog; DNA repair exonuclease REC1; exonuclease homolog RAD1; hRAD1; HRAD1 REC1 DNA repair exonuclease REC1 DNA repair exonuclease rad1 homolog cell cycle checkpoint protein Hrad1 cell cycle checkpoint protein RAD1 checkpoint control protein HRAD1 exonuclease homolog RAD1 rad1-like DNA damage checkpoint protein; RAD1 checkpoint clamp component; RAD1 homolog; Rad1-like DNA damage checkpoint protein
Gene Aliases: HRAD1; RAD1; REC1
UniProt ID: (Human) O60671
Entrez Gene ID: (Human) 5810
Molecular Function: DNA binding protein exodeoxyribonuclease hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.