Invitrogen
Manufacturer Code:PA5114775
Catalog # PIPA5114775
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RAD1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cell cycle checkpoint protein Hrad1; cell cycle checkpoint protein Hrad1 Cell cycle checkpoint protein RAD1 checkpoint control protein HRAD1 DNA repair exonuclease rad1 homolog DNA repair exonuclease REC1 exonuclease homolog RAD1 hRAD1 mRAD1 Rad1 RAD1 checkpoint clamp component RAD1 checkpoint DNA exonuclease RAD1 homolog RAD1 homolog (S. pombe) rad1-like DNA damage checkpoint protein Rec1; Cell cycle checkpoint protein RAD1; checkpoint control protein HRAD1; DNA repair exonuclease rad1 homolog; DNA repair exonuclease REC1; exonuclease homolog RAD1; hRAD1; mRAD1; RAD1 checkpoint clamp component; RAD1 homolog; Rad1-like DNA damage checkpoint protein
Gene Aliases: HRAD1; mRAD1; RAD1; REC1
UniProt ID: (Human) O60671, (Mouse) Q9QWZ1
Entrez Gene ID: (Human) 5810, (Mouse) 19355
Molecular Function: DNA binding protein exodeoxyribonuclease hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.