Invitrogen
Manufacturer Code:PA554282
Catalog # PIPA554282
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RACK7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTCL tumor antigen se14-3; CTCL-associated antigen se14-3; Cutaneous T-cell lymphoma-associated antigen se14-3; predicted protein of HQ2893; PRKCBP1 RACK7 PRO2893 zinc finger MYND-type containing 8 ZMYND8; protein kinase C binding protein 1; Protein kinase C-binding protein 1; Rack7; spikar delta C; Zinc finger MYND domain-containing protein 8; zinc finger, MYND-type containing 8
Gene Aliases: 1110013E22Rik; 2010005I16Rik; 3632413B07Rik; AI316811; AL024039; KIAA1125; PRKCBP1; PRO2893; RACK7; spikar; ZMYND8
UniProt ID: (Human) Q9ULU4
Entrez Gene ID: (Human) 23613, (Mouse) 228880, (Rat) 296374
Molecular Function: ligase transcription cofactor transcription factor ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.