Novus Biologicals
Manufacturer Code:NBP273781JF646
Catalog # N273781J646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D8 |
Immunogen | Full length recombinant protein of human RACK1/GNB2L1 (NP_006089) produced in HEK293T cells. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Cell proliferation-inducing gene 21 protein; guanine nucleotide binding guanine nucleotide binding protein (G protein) beta polypeptide 2-like 1 Receptor for activated C kinase Receptor for Activated C Kinase 1; guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1; guanine nucleotide binding protein beta polypeptide 2-like 1; Guanine nucleotide-binding protein subunit beta-2-like 1; Guanine nucleotide-binding protein subunit beta-2-like 1, N-terminally processed; Guanine nucleotide-binding protein subunit beta-like protein 12.3; HLC-7; Human lung cancer oncogene 7 protein; lung cancer oncogene 7; proliferation-inducing gene 21; protein homologous to chicken B complex protein, guanine nucleotide binding; Receptor for activated C kinase; Receptor of activated protein C kinase 1; Receptor of activated protein C kinase 1, N-terminally processed; receptor of activated protein kinase C 1; Small ribosomal subunit protein RACK1
Gene Aliases: Gnb2-rs1; GNB2L1; H12.3; HLC-7; HLC7; PIG21; RACK1
UniProt ID: (Human) P63244
Entrez Gene ID: (Human) 10399
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.