Invitrogen
Manufacturer Code:MA534809
Catalog # PIMA534809
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG40-26 |
Immunogen | Recombinant protein corresponding to C-terminal human RACK1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 12-3 AL033335 Cell proliferation-inducing gene 21 protein CG7111 DmelCG7111 Dmel_CG7111 dRack1 GB-like GNB2L1 Gnb2-rs1 Guanine nucleotide-binding protein subunit beta-2-like 1 Guanine nucleotide-binding protein subunit beta-2-like 1 N-terminally processed Guanine nucleotide-binding protein subunit beta-like protein Guanine nucleotide-binding protein subunit beta-like protein 12.3 H12.3 HLC7 HLC-7 Human lung cancer oncogene 7 protein i173 OTTHUMP00000223870 OTTHUMP00000223891 OTTHUMP00000223893 OTTHUMP00000223900 OTTHUMP00000223901 OTTHUMP00000223902 OTTHUMP00000223930 OTTHUMP00000223931 p205 PIG21 RACK rack1 Receptor for activated C kinase Receptor of activated protein C kinase 1 Receptor of activated protein C kinase 1 N-terminally processed Receptor of activated protein kinase C Receptor of activated protein kinase C 1 Receptor of activated protein kinase C homolog Small ribosomal subunit protein RACK1 wu:fb80d08 wu:fk65d12; Cell proliferation-inducing gene 21 protein; guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1; guanine nucleotide binding protein beta polypeptide 2-like 1; Guanine nucleotide-binding protein subunit beta-2-like 1; Guanine nucleotide-binding protein subunit beta-2-like 1, N-terminally processed; Guanine nucleotide-binding protein subunit beta-like protein 12.3; HLC-7; Human lung cancer oncogene 7 protein; lung cancer oncogene 7; proliferation-inducing gene 21; protein homologous to chicken B complex protein, guanine nucleotide binding; Receptor for activated C kinase; Receptor of activated protein C kinase 1; Receptor of activated protein C kinase 1, N-terminally processed; receptor of activated protein kinase C 1; Small ribosomal subunit protein RACK1
Gene Aliases: Gnb2-rs1; GNB2L1; H12.3; HLC-7; HLC7; PIG21; RACK1
UniProt ID: (Human) P63244
Entrez Gene ID: (Human) 10399
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.