Invitrogen
Manufacturer Code:MA535405
Catalog # PIMA535405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0837 |
Immunogen | A synthesized peptide derived from human RACK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 12-3; 12-3 AL033335 Cell proliferation-inducing gene 21 protein CG7111 DmelCG7111 Dmel_CG7111 dRack1 GB-like GNB2L1 Gnb2-rs1 Guanine nucleotide-binding protein subunit beta-2-like 1 Guanine nucleotide-binding protein subunit beta-2-like 1 N-terminally processed Guanine nucleotide-binding protein subunit beta-like protein Guanine nucleotide-binding protein subunit beta-like protein 12.3 H12.3 HLC7 HLC-7 Human lung cancer oncogene 7 protein i173 OTTHUMP00000223870 OTTHUMP00000223891 OTTHUMP00000223893 OTTHUMP00000223900 OTTHUMP00000223901 OTTHUMP00000223902 OTTHUMP00000223930 OTTHUMP00000223931 p205 PIG21 RACK rack1 Receptor for activated C kinase Receptor of activated protein C kinase 1 Receptor of activated protein C kinase 1 N-terminally processed Receptor of activated protein kinase C Receptor of activated protein kinase C 1 Receptor of activated protein kinase C homolog Small ribosomal subunit protein RACK1 wu:fb80d08 wu:fk65d12; activated protein kinase C receptor; Cell proliferation-inducing gene 21 protein; guanine nucleotide binding protein (G protein) beta polypeptide 2-like 1; guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1 sequence 1; guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1; guanine nucleotide binding protein beta polypeptide 2-like 1; guanine nucleotide binding protein related; guanine nucleotide binding protein, beta 2, related sequence 1; guanine nucleotide binding protein, beta polypeptide 2-like 1; guanine nucleotide binding protein, beta-2, related sequence 1; Guanine nucleotide-binding protein subunit beta-2-like 1; Guanine nucleotide-binding protein subunit beta-2-like 1, N-terminally processed; Guanine nucleotide-binding protein subunit beta-like protein 12.3; HLC-7; Human lung cancer oncogene 7 protein; lung cancer oncogene 7; p205; proliferation-inducing gene 21; protein homologous to chicken B complex protein, guanine nucleotide binding; protein kinase C receptor; Receptor for activated C kinase; receptor for activated protein kinase C; Receptor of activated protein C kinase 1; Receptor of activated protein C kinase 1, N-terminally processed; Receptor of activated protein kinase C 1; Small ribosomal subunit protein RACK1
Gene Aliases: AL033335; GB-like; Gnb2-rs1; GNB2L1; H12.3; HLC-7; HLC7; p205; PIG21; RACK1
UniProt ID: (Rat) P63245, (Mouse) P68040, (Human) P63244
Entrez Gene ID: (Rat) 83427, (Mouse) 14694, (Human) 10399
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.