Invitrogen
Manufacturer Code:PA131510
Catalog # PIPA131510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence GRQGNFFASPMLK from C Terminus of the protein sequence according to NP&lowbar037409 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CYK4; HsCYK-4 ID; Male germ cell RacGap; MgcRacGAP; Protein CYK4 homolog; Rac GTPase-activating protein 1
Gene Aliases: CYK4; HsCYK-4; ID-GAP; KIAA1478; MGCRACGAP; RACGAP1
UniProt ID: (Human) Q9H0H5
Entrez Gene ID: (Human) 29127
Molecular Function:
G-protein modulator
enzyme modulator
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.