Abnova
Manufacturer Code:H00005880A01
Catalog # 89-003-064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | RAC2 (AAH01485.1 1 a.a. ~ 192 a.a) full-length recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length recombinant RAC2.
For Research Use Only
Protein Aliases: EN-7; GX; HSPC022; p21-Rac2; Ras-related C3 botulinum toxin substrate 2; Ras-related C3 botulinum toxin substrate 3 (rho family, small GTP-binding protein Rac2); Small G protein
Gene Aliases: EN-7; Gx; HSPC022; p21-Rac2; RAC2
UniProt ID: (Human) P15153
Entrez Gene ID: (Human) 5880
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.