Proteintech
Manufacturer Code:119161AP150UL
Catalog # 50-172-7557
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RABL3 Fusion Protein Ag2531 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Rab like protein 3 RABL3; RAB, member of RAS oncogene family-like 3; Rab-like protein 3
Gene Aliases: 4930553C05Rik; AA958834; RABL3
UniProt ID: (Human) Q5HYI8, (Mouse) Q9D4V7
Entrez Gene ID: (Human) 285282, (Mouse) 67657, (Rat) 360720
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.