Invitrogen
Manufacturer Code:PA543203
Catalog # PIPA543203
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human RABGGTB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Geranylgeranyl transferase type II subunit beta; Geranylgeranyl transferase type-2 subunit beta; GGTase-II-beta; Rab geranyl-geranyltransferase subunit beta; Rab geranylgeranyl transferase componenet, subunit beta; Rab geranylgeranyltransferase beta subunit; Rab geranylgeranyltransferase subunit beta; RAB geranylgeranyltransferase, beta subunit; Rab GG transferase beta; rab GGTase beta; Type II protein geranyl-geranyltransferase subunit beta
Gene Aliases: fj12b07; GGTB; RABGGTB; wu:fj12b07; zgc:56443
UniProt ID: (Human) P53611, (Bovine) Q5E9B3, (Mouse) P53612, (Rat) Q08603
Entrez Gene ID: (Dog) 612683, (Horse) 100067295, (Sheep) 100302071, (Human) 5876, (Rabbit) 100345858, (Bovine) 533276, (Guinea pig) 100715556, (Mouse) 19352, (Rat) 25533, (Zebrafish) 406386
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.