Invitrogen
Manufacturer Code:PA528324
Catalog # PIPA528324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 37 and 318 of Human RABGGTB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Geranylgeranyl transferase type II subunit beta; Geranylgeranyl transferase type-2 subunit beta; GGTase-II-beta; GGTB GGTase-II-beta geranylgeranyl transferase type II subunit beta geranylgeranyl transferase type-2 subunit beta rab GG transferase beta rab GGTase beta rab geranyl-geranyltransferase subunit beta rab geranylgeranyltransferase subunit beta type II protein ge; Rab geranyl-geranyltransferase subunit beta; Rab geranylgeranyl transferase componenet, subunit beta; Rab geranylgeranyltransferase subunit beta; RAB geranylgeranyltransferase, beta subunit; Rab GG transferase beta; rab GGTase beta; Type II protein geranyl-geranyltransferase subunit beta
Gene Aliases: GGTB; RABGGTB
UniProt ID: (Human) P53611, (Mouse) P53612, (Rat) Q08603
Entrez Gene ID: (Human) 5876, (Mouse) 19352, (Rat) 25533
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.