Novus Biologicals
Manufacturer Code:NBP232214
Catalog # NBP232214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 281 to 331 of human Rab Geranylgeranyltransferase Beta Subunit using the numbering given in entry NP_004573.2 (GeneID 5876). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.5.1.60 Geranylgeranyl transferase type II subunit beta geranylgeranyl transferase type-2 subunit beta GGTase-II-beta GGTB Rab geranylgeranyltransferase subunit beta Rab geranyl-geranyltransferase subunit beta Rab geranylgeranyltransferase beta subunit Rab GG transferase beta Rab GGTase beta Type II protein geranyl-geranyltransferase subunit beta; Geranylgeranyl transferase type II subunit beta; Geranylgeranyl transferase type-2 subunit beta; GGTase-II-beta; Rab geranyl-geranyltransferase subunit beta; Rab geranylgeranyltransferase subunit beta; Rab geranylgeranyltransferase, beta subunit; Rab GG transferase beta; rab GGTase beta; Type II protein geranyl-geranyltransferase subunit beta
Gene Aliases: GGTB; RABGGTB
UniProt ID: (Human) P53611
Entrez Gene ID: (Human) 5876
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.