Invitrogen
Manufacturer Code:PA569572
Catalog # PIPA569572
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human RABGGTA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Geranylgeranyl transferase type II subunit alpha; Geranylgeranyl transferase type-2 subunit alpha; gunmetal; protein prenyltransferase alpha subunit repeat containing 3; Rab geranyl-geranyltransferase subunit alpha; Rab geranylgeranyl transferase, a subunit; Rab geranylgeranyltransferase alpha; Rab geranylgeranyltransferase alpha subunit; Rab geranylgeranyltransferase subunit alpha; Rab geranylgeranyltransferase, alpha subunit; Rab GG transferase alpha; Rab GGTase alpha
Gene Aliases: Ggta; gm; PTAR3; RABGGTA
UniProt ID: (Bovine) Q5EA80, (Human) Q92696, (Mouse) Q9JHK4, (Rat) Q08602
Entrez Gene ID: (Horse) 100051161, (Bovine) 516619, (Human) 5875, (Dog) 608407, (Rabbit) 100349944, (Guinea pig) 100735569, (Mouse) 56187, (Rat) 58983
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.