Proteintech
Manufacturer Code:138941AP150UL
Catalog # 50-172-9072
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RABGAP1L Fusion Protein Ag4856 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: expressed in hematopoietic cells, heart, liver (HLL); HHL KIAA0471 RABGAP1L TBC1D18; RAB GTPase activating protein 1-like; Rab GTPase-activating protein 1-like; Rab GTPase-activating protein 1-like, isoform 10; rab6 GTPase activating protein-like; RABGAP1L; TBC1 domain family, member 18
Gene Aliases: 5830411O09Rik; 8430421H08Rik; 9630005B12Rik; AW049894; Hh1; HHL; KIAA0471; mKIAA0471; RABGAP1L; RGD1304620; TBC1D18
UniProt ID: (Human) B7ZAP0, (Mouse) A6H6A9
Entrez Gene ID: (Human) 9910, (Mouse) 29809, (Rat) 304914
Molecular Function:
G-protein modulator
enzyme modulator
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.