Proteintech
Manufacturer Code:151051AP150UL
Catalog # 50-172-9982
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RABEPK/p40 Fusion Protein Ag2593 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 40 kDa Rab9 effector protein; 40 kDa Rab9 effector protein p40 RAB9P40 RABEPK; p40; Rab9 effector p40; Rab9 effector protein with kelch motifs
Gene Aliases: bA65N13.1; p40; RAB9P40; RABEPK
UniProt ID: (Human) Q7Z6M1
Entrez Gene ID: (Human) 10244
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.