Proteintech
Manufacturer Code:102132AP150UL
Catalog # 50-172-6118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RABEPK/p40 Fusion Protein Ag0285 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 40 kDa Rab9 effector protein; 40 kDa Rab9 effector protein p40 RAB9P40 RABEPK; p40; Rab9 effector p40; Rab9 effector protein with kelch motifs
Gene Aliases: 8430412M01Rik; 9530020D24Rik; AV073337; bA65N13.1; C87311; p40; RAB9P40; RABEPK; RGD1310612
UniProt ID: (Human) Q7Z6M1, (Rat) Q4V8F4, (Mouse) Q8VCH5
Entrez Gene ID: (Human) 10244, (Rat) 296649, (Mouse) 227746
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.