Invitrogen
Manufacturer Code:PA545012
Catalog # PIPA545012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human RAB8B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTPase Rab8b; RAB-8b protein; RAB44, member RAS oncogene family; RAB8B member RAS oncogene family FLJ38125; Ras-related protein Rab-8B; Ras-related protein Rab-8B-like protein
Gene Aliases: 5930437D16; D330025I23Rik; RAB8B; rab8bb; zgc:165529
UniProt ID: (Bovine) Q2HJI8, (Human) Q92930, (Rat) P70550, (Mouse) P61028
Entrez Gene ID: (Goat) 102172357, (Sheep) 101111969, (Bovine) 614242, (Dog) 610601, (Horse) 100054073, (Rabbit) 100349093, (Human) 51762, (Rat) 266688, (Mouse) 235442, (Guinea pig) 100727085, (Zebrafish) 100093706
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.