Invitrogen
Manufacturer Code:PA537209
Catalog # PIPA537209
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to residues in human RAB8B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTPase Rab8b; RAB-8b protein; RAB44, member RAS oncogene family; Ras-related protein Rab-8B
Gene Aliases: 5930437D16; D330025I23Rik; RAB8B
UniProt ID: (Human) Q92930, (Rat) P70550, (Mouse) P61028
Entrez Gene ID: (Human) 51762, (Rat) 266688, (Mouse) 235442
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.