Invitrogen
Manufacturer Code:PA5104086
Catalog # PIPA5104086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human RAB9B corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5930437D16 D330025I23Rik FLJ38125 GTPase Rab8b RAB44 member RAS oncogene family Rab8b RAB-8b protein RAB8B member RAS oncogene family ras-related protein Rab-8B; GTPase Rab8b; RAB-8b protein; RAB44, member RAS oncogene family; Ras-related protein Rab-8B
Gene Aliases: 5930437D16; D330025I23Rik; RAB8B
UniProt ID: (Human) Q92930, (Rat) P70550, (Mouse) P61028
Entrez Gene ID: (Human) 51762, (Rat) 266688, (Mouse) 235442
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.