Invitrogen
Manufacturer Code:MA524717
Catalog # PIMA524717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 261CT1.3.1 |
Immunogen | human RAB8A recombit protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cell line NK14 derived transforming oncogene; mel transforming oncogene (derived from cell line NK14); mel transforming oncogene (derived from cell line NK14)- RAB8 homolog; mel transforming oncogene (RAB8 homolog); Oncogene c-mel; ras-associated protein RAB8; Ras-related protein Rab-8A; Ras-related protein Rab-8A Oncogene c-mel RAB8A MEL RAB8
Gene Aliases: AA409338; MEL; RAB8; RAB8A
UniProt ID: (Human) P61006, (Mouse) P55258
Entrez Gene ID: (Human) 4218, (Mouse) 17274
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.